Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-1-(7-chloro-2,2-bis(fluoromethyl)chroman-4-yl)-3-(3-methylisoquinolin-5-yl)urea | 1221443-94-2 | 99.9% | 431.86 | C22H20ClF2N3O2 | 10MM 1ML
SDP

Supplier:  Medchemexpress LLC HY1242810MM 1ML

Encompass_Preferred

A-1165442 is a potent, temperature-neutral TRPV1 antagonist described in the medicinal chemistry literature and used for preclinical analgesia research. It exhibits high potency against human TRPV1 and is supplied as a ready-to-use 10 mM solution in DMSO intended for in vitro and pharmacology studies.

  • Potent TRPV1 antagonism (IC50 ≈ 9 nM for human TRPV1).
  • Temperature-neutral profile with demonstrated analgesic efficacy in preclinical models.
  • Supplied as 10 mM solution in DMSO, 1 mL vial, ready for use.
  • Vendor-reported purity 99.92%.
  • Molecular weight 431.86 g/mol; formula C22H20ClF2N3O2.
  • Recommended storage: powder -20°C; in solvent -80°C for long-term stability.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-37891


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.