Learn More
Medchemexpress LLC (R)-1-Amino-18-(3,4-bis((4-fluorobenzyl)oxy)phenoxy)-3,6,9,12-tetraoxa-15-azaoctadecan-17-ol | 99.5% | 634.71 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16938750MG
Encompass_Preferred
YT 6-2-PEG3-C2-NH2 is an autophagy-targeting ligand-linker conjugate. It is designed to target p62/SQSTM1 and is associated with AUTOTAC ATC-324, a bivalent AR (Androgen Receptor) degrader. This product can reduce nuclear AR levels, downregulate AR and AR-v7 target gene expression, and degrade common AR mutants in PCa.
  • Appearance as a colorless to light yellow oil
  • Consistent with structure by 1H NMR spectrum and LCMS
  • High purity confirmed by HPLC
  • Suitable for research applications
Catalog No. 50-003-90830