Learn More
Medchemexpress LLC (R)-1-benzylpyrrolidine-2-carboxylic acid | 56080-99-0 | 99.0% | 205.25 | C12H15NO2 | 100 G

Supplier: Medchemexpress LLC HYW061650100G
(R)-1-Benzylpyrrolidine-2-carboxylic acid is an R-enantiomer proline derivative used as a research reagent and building block in organic synthesis and biochemical studies. The compound is supplied with analytical characterization and safety documents for research use only.
- Purity 98.99%.
- Chemical formula C12H15NO2.
- Molecular weight 205.25 g·mol⁻¹.
- Available sizes 10 g, 25 g, 100 g.
- Includes data sheet, COA, SDS, HNMR, and RP-HPLC files.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.