Learn More
Cayman Chemical R-1 MethndamIde 10mg
SDP
Supplier:  Cayman Chemical 9007010 MG
Encompass_Preferred
A potent CB1 receptor agonist in the methanandamide series; selective for CB1 over CB2 receptors (Kis = 17.9-28.9 and 815-868 nM, respectively); more potent than AEA and more resistant than AEA to hydrolytic inactivation by FAAH
Catalog No. 50-289-5854