Learn More
Strem, An Ascensus Company CAS# 1365531-93-6. 100mg. (R)-2,2'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxy)-1,1'-biphenyl, min. 97% (R)-DMM-Garphos™. MFCD19443626
SDP
Supplier:  Strem, An Ascensus Company 151666100MG
Encompass
CAS# 1365531-93-6. 100mg. (R)-2,2'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxy)-1,1'-biphenyl, min. 97% (R)-DMM-Garphos™. MFCD19443626. Molecular Weight: 874.98. Molecular Formula: C52H60O8P2. Color/form: white xtl. Strem# 15-1666. http://www.strem.com/catalog/v/15-1666/
Catalog No. 50-370-8