Learn More
Strem, An Ascensus Company (R)-MeO-BIPHEP min. 97% 500mg
SDP
Supplier:  Strem, An Ascensus Company 150178500MG
Encompass
CAS# 133545-16-1. 500mg. (R)-(+)-2,2'-Bis(diphenylphosphino)-6,6'-dimethoxy-1,1'-biphenyl, min. 97% (R)-MeO-BIPHEP. MFCD03095430. Molecular Weight: 582.53. Molecular Formula: C38H32O2P2. Color/form: off-white xtl. Strem# 15-0178. http://www.strem.com/catalog/v/15-0178/
Catalog No. 50-901-09827