Learn More
Medchemexpress LLC (R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl | 76189-55-4 | MFCD00010805 | 98.0% | 622.67 g·mol⁻¹ | C44H32P2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01775710G
Encompass_Preferred
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl is a chiral bisphosphine ligand supplied as a solid research reagent for use in asymmetric synthesis and transition-metal catalysis. It is provided for laboratory research use only and typically appears as a white to pale-yellow crystalline powder suitable for weighing and handling in small-scale reactions.
  • Chiral bisphosphine ligand for asymmetric catalysis.
  • High reported purity suitable for research applications.
  • Solid, crystalline powder, easy to weigh and handle.
  • Compatible with common transition-metal catalysts such as palladium and rhodium.
  • Used to induce enantioselectivity in organic synthesis.
  • Available in small pack sizes for laboratory use.
Catalog No. 50-002-99913