Learn More
Strem, An Ascensus Company CAS# 133545-24-1. 500mg. (R)-(+)-2,2'-Bis(di-p-tolylphosphino)-6,6'-dimethoxy-1,1'-biphenyl, min. 97%. MFCD09753006
SDP
Supplier:  Strem, An Ascensus Company 150156500MG
Encompass
CAS# 133545-24-1. 500mg. (R)-(+)-2,2'-Bis(di-p-tolylphosphino)-6,6'-dimethoxy-1,1'-biphenyl, min. 97%. MFCD09753006. Molecular Weight: 638.73. Molecular Formula: C42H40O2P2. Color/form: white to pale yellow solid Strem# 15-0156. http://www.strem.com/catalog/v/15-0156/
Catalog No. 50-138-5358