Learn More
Medchemexpress LLC (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(thiophen-3-yl)propanoic acid | 220497-90-5 | 99.4% | 393.46 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW011003
Encompass_Preferred
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(thiophen-3-yl)propanoic acid is a chemical compound primarily used in laboratory settings for research and the manufacture of various substances.
  • Used for laboratory research and substance manufacturing.
  • Appears as a white to off-white solid.
  • Has a molecular formula of C22H19NO4S and a molecular weight of 393.46.
  • Purity is 99.35% by LCMS.
  • Store as powder at -20°C for up to 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.
  • Spectra consistent with chemical structure.
Catalog No. 50-004-53333