Learn More
STA PHARMACEUTICAL US LLC (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)undecanoic acid | 1 g | CAS 1821771-29-2 | InChIKey IGRGFQSURTWTSB-XMMPIXPASA-N
Supplier: STA PHARMACEUTICAL US LLC STAAA102591GM
(R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)undecanoic acid is a Amino Acid reagent (Subcategory: Lipidated AA) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 1821771-29-2
- MDL: No data
- InChIKey: IGRGFQSURTWTSB-XMMPIXPASA-N
- Molecular Weight: 423.553
- Molecular Formula: C26H33NO4
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)undecanoic acid
- SMILES: CCCCCCCCC[C@H](C(O)=O)NC(OCC1C2=CC=CC=C2C3=CC=CC=C13)=O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.