Learn More
Medchemexpress LLC (R)-2-amino-2-(thiophen-3-yl)acetic acid | 1194-86-1 | MFCD00079614 | 98.0% | 157.19 g/mol | C6H7NO2S | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0172545G
Encompass_Preferred
(R)-2-Amino-2-(thiophen-3-yl)acetic acid is the R-enantiomer of a thiophene-substituted amino acid used as a research reagent and synthetic building block in medicinal chemistry and peptide synthesis. The compound is supplied as a white to off-white solid with a molecular formula of C6H7NO2S, molecular weight 157.19 g/mol, and a typical purity of ≥98.0%.
  • R-enantiomer for stereospecific synthesis or research applications.
  • High purity suitable for analytical and synthetic use.
  • Solid, white to off-white appearance for easy handling.
  • Available in multiple pack sizes for scale flexibility.
  • Useful as a building block in peptide and small-molecule synthesis.
Catalog No. 50-002-99907