Learn More
Medchemexpress LLC (R)-2-amino-3-(3-(methylsulfonyl)phenyl)propanoic acid | 1270093-99-6 | ≥97.0% | 243.28 | 1 G
SDP
Supplier:  Medchemexpress LLC HY235291G
Encompass_Preferred
Lifitegrast impurity 6 is identified as an impurity of Lifitegrast and serves as a drug intermediate. It is supplied as a solid, white to off-white in appearance.
  • Impurity of Lifitegrast
  • Functions as a drug intermediate
  • Solid, white to off-white appearance
  • Recommended storage at room temperature for 3 years, or in solvent at -80°C for 2 years or -20°C for 1 year
  • For research use only, not fully validated for medical applications
Catalog No. 50-004-00880