Learn More
Medchemexpress LLC (R)-2-(((Benzyloxy)carbonyl)amino)-3,3-dimethylbutanoic acid | 70874-05-4 | 96.2% | C14H19NO4 | 1 G

Supplier: Medchemexpress LLC HY776301G
(R)-2-(((Benzyloxy)carbonyl)amino)-3,3-dimethylbutanoic acid is a chemical compound characterized by its molecular formula C₁₄H₁₉NO₄ and a molecular weight of 265.30. It presents as a colorless to light yellow liquid with a purity of 96.17% via HPLC. This product is for research use only and has not been fully validated for medical applications.
- Molecular formula: C₁₄H₁₉NO₄
- Molecular weight: 265.30
- High purity of 96.17% (HPLC)
- Appearance: Colorless to light yellow liquid
- Stable for 3 years at -20°C in pure form
- Stable for 2 years at 4°C in pure form
- Stable for 6 months at -80°C in solvent
- Stable for 1 month at -20°C in solvent
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.