Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-2-(((Benzyloxy)carbonyl)amino)-3,3-dimethylbutanoic acid | 70874-05-4 | 96.2% | C14H19NO4 | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHY77630

Encompass_Preferred

This product is (R)-2-(((Benzyloxy)carbonyl)amino)-3,3-dimethylbutanoic acid, a chemical compound from MedChemExpress. It has a molecular formula of C14H19NO4 and a molecular weight of 265.30.

  • Purity of 96.17% by HPLC
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Appearance as a colorless to light yellow liquid

Catalog No. 50-003-58609


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.