Learn More
STA PHARMACEUTICAL US LLC (R)-2-(Fmoc-amino)heptanoic acid | 10 g | CAS 1629051-80-4 | MDL MFCD08275893
Supplier: STA PHARMACEUTICAL US LLC STAAA1136210GM
(R)-2-(Fmoc-amino)heptanoic acid is a Amino Acid reagent (Subcategory: Lipidated AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 1629051-80-4
- MDL: MFCD08275893
- InChIKey: DSEDZIUESKRBOW-HXUWFJFHSA-N
- Molecular Weight: 367.445
- Molecular Formula: C22H25NO4
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)heptanoic acid
- SMILES: CCCCC[C@H](C(O)=O)NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.