Learn More
Medchemexpress LLC (R)-2-hydroxy-2-methyl-4-(2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl)butanamide | 1147883-03-1 | 99.3% | 265.30 g/mol | C14H19NO4 | 10MG

Supplier: Medchemexpress LLC HY12599910MG
EPI-589 is a redox-active neuroprotectant under investigation for neurodegenerative disease research; it acts as an oxidoreductase inhibitor and free-radical scavenger with demonstrated blood-brain barrier permeability and oral activity in preclinical studies. The compound is supplied as a light-yellow solid with high reported purity and common small-package options suitable for in vitro and in vivo research.
- Redox-active neuroprotectant for research use.
- Reversible redox cofactor and free-radical scavenger.
- High purity (99.3%).
- Soluble in DMSO at 100 mg/mL (use freshly opened DMSO; ultrasonic assistance may be needed).
- Available in small package sizes suitable for early-stage studies.
- Solid, light yellow appearance facilitating handling and storage.
- Recommended storage: powder at -20 °C (up to 3 years) or 4 °C (up to 2 years); in solution at -80 °C (up to 6 months) or -20 °C (up to 1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.