Learn More
Medchemexpress LLC (R)-3-(1H-indol-3-yl)-2-(5-(p-tolylethynyl)thiophene-2-sulfonamido)propanoic acid | 203640-27-1 | MFCD22200275 | 99.8% | 464.56 | C24H20N2O4S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY106992100MG
Encompass_Preferred
S 3304 is a research-grade small-molecule inhibitor of matrix metalloproteinases with demonstrated selectivity for MMP-2 and MMP-9. The compound contains an alkyne functional group enabling click-chemistry applications and is formulated as a high-purity solid for use in in vitro and in vivo studies.
  • Selective inhibition of MMP-2 and MMP-9.
  • Contains an alkyne moiety compatible with copper-catalyzed azide-alkyne cycloaddition.
  • Supplied as a light yellow solid with high purity.
  • High solubility in DMSO for in vitro applications.
  • Protocols available for in vivo formulation and dosing.
  • Stable under frozen storage to preserve activity.
  • Provided in a 100 mg research package.
Catalog No. 50-002-39979