Learn More
Medchemexpress LLC (R)-3-(1H-indol-3-yl)-2-(5-(p-tolylethynyl)thiophene-2-sulfonamido)propanoic acid | 203640-27-1 | MFCD22200275 | 99.8% | 464.56 g/mol | C24H20N2O4S2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10699210MG
Encompass_Preferred
S 3304 is a research reagent that inhibits matrix metalloproteinases MMP-2 and MMP-9. It is the (R)-3-(1H-indol-3-yl)-2-(5-(p-tolylethynyl)thiophene-2-sulfonamido)propanoic acid (CAS 203640-27-1), formula C24H20N2O4S2, molecular weight 464.56 g/mol. The compound is supplied as a light yellow solid and is available in solution formats for laboratory research use only.
  • Selective inhibitor of MMP-2 and MMP-9.
  • Reported to inhibit angiogenesis and metastasis in vivo.
  • High purity, typically 99.75%.
  • Available as solid and as 10 mM DMSO solution formats.
  • Provided with certificate of analysis and safety data sheet.
Catalog No. 50-002-40006