Learn More
Medchemexpress LLC (R)-3-hydroxytetradecanoic acid | 28715-21-1 | MFCD00211333 | >=98.0% | 244.37 g/mol | C14H28O3 | 5 MG

Supplier: Medchemexpress LLC HYW2883755MG
(R)-3-Hydroxytetradecanoic acid is the R-enantiomer of a 14-carbon hydroxy fatty acid used as a research reagent in biochemical and metabolic studies. It has defined stereochemistry, confirmed purity by analytical methods, and is supplied in small quantities for laboratory use.
- Chiral C14 hydroxy fatty acid, R-enantiomer.
- High purity confirmed by NMR (≥98.0%).
- Molecular formula C14H28O3; molecular weight 244.37 g/mol.
- Suitable for biochemical and metabolic research applications.
- Storage: room temperature for long term; in solvent, -80 °C or -20 °C as specified.
- Provided in small research quantities (5 mg).
- CAS number 28715-21-1 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.