Learn More
Medchemexpress LLC (R)-4-Benzyl-2-oxazolidinone | 102029-44-7 | 177.20 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW007664100G
Encompass_Preferred
(R)-4-Benzyl-2-oxazolidinone is an oxazolidinone-type auxiliary that enables highly stereoselective asymmetric alkylation reactions. It is applied in the total synthesis of natural products and pharmaceutical preparations. In pharmaceutical research and development, (R)-4-Benzyl-2-oxazolidinone serves as a key precursor for the synthesis of aromatase inhibitors and can be used in studies related to estrogen-dependent breast cancer.
  • (R)-4-Benzyl-2-oxazolidinone serves as the core chiral parent nucleus of aromatase inhibitors, and the inhibitory activity of derivatives of (R)-4-Benzyl-2-oxazolidinone is significantly superior to that of (S)-4-Benzyl-2-oxazolidinone.
Catalog No. 50-004-48651