Learn More
Medchemexpress LLC (R)-4-Benzyl-2-oxazolidinone | 102029-44-7 | 177.20 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW007664
Encompass_Preferred
(R)-4-Benzyl-2-oxazolidinone is an oxazolidinone-type auxiliary that facilitates highly stereoselective asymmetric alkylation reactions. It is utilized in the total synthesis of natural products and in various pharmaceutical preparations. This compound also serves as a key precursor for the synthesis of aromatase inhibitors and is relevant in studies concerning estrogen-dependent breast cancer.
  • Enables highly stereoselective asymmetric alkylation reactions.
  • Applied in the total synthesis of natural products.
  • Used in pharmaceutical preparations.
  • Key precursor for the synthesis of aromatase inhibitors.
  • Relevant for studies related to estrogen-dependent breast cancer.
Catalog No. 50-004-49321