Learn More
Electron Microscopy Sciences MohrPro R-401 Dev/Activator/Cleaner 1 QT
SDP
Supplier:  Electron Microscopy Sciences 7496501
Encompass_Preferred
All chemicals are concentrated . One quart (0.95L) of concentrate mixed with water will make one gallon of working solution.
Catalog No. 50-249-56