Learn More
Medchemexpress LLC (R)-5-((2,3-difluorobenzyl)thio)-7-((1-hydroxypropan-2-yl)amino)thiazolo[4,5-d]pyrimidin-2(3H)-one | 333742-48-6 | 100.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1192595MG
Encompass_Preferred
AZD8309 is an orally active small-molecule antagonist of the C-X-C chemokine receptor 2 (CXCR2), used as a research tool to modulate neutrophil transmigration and investigate inflammatory disease mechanisms.
  • Orally active CXCR2 antagonist for in vitro and in vivo studies.
  • Useful for studying neutrophil transmigration and inflammation pathways.
  • Available in small research quantities (5 mg solid) and DMSO solution formats.
  • Molecular weight 384.42 g·mol⁻¹; molecular formula C15H14F2N4O2S2.
  • Provided as a research-grade compound for laboratory use only.
Catalog No. 50-002-34667