Learn More
Medchemexpress LLC (R)-5,7-difluorochroman-4-ol | 1270294-05-7 | MFCD09763634 | 100.0% | 186.16 g/mol | C9H8F2O2 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW048541A100G
Encompass_Preferred
(R)-5,7-Difluorochroman-4-ol is a chiral drug intermediate used in medicinal chemistry and synthetic research for the preparation of pharmaceutically active compounds. It is supplied as a high-purity enantiomer suitable for laboratory synthesis and analytical work.
  • High purity suitable for synthetic applications and analytical work.
  • Molecular formula C9H8F2O2 and molecular weight 186.16 g/mol.
  • Available in multiple pack sizes including 100 G for scale-up or bulk use.
  • Commonly used as an intermediate in the synthesis of drug candidates.
  • Supplied as the (R)-enantiomer for stereoselective synthesis.
Catalog No. 50-002-10874