Learn More
Medchemexpress LLC (R)-5-Oxopyrrolidine-2-carboxylic acid | 4042-36-8 | 129.11 | 1 ML

Supplier: Medchemexpress LLC HYW00230410MM/1ML
(R)-5-Oxopyrrolidine-2-carboxylic acid, also known as D-Pyroglutamic acid, is a cyclic derivative of D-glutamate. It is used in research studies related to metabolic syndrome and end-stage renal disease.
- Cyclic derivative of D-glutamate
- Used in metabolic syndrome research
- Used in end-stage renal disease research
- Purity of 98.0%
- Molecular weight of 129.11
- Formula is C5H7NO3
- Appears as a white to off-white solid
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.