Learn More
Medchemexpress LLC (R)-5-Oxopyrrolidine-2-carboxylic acid | 4042-36-8 | 129.11 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW002304
Encompass_Preferred
(R)-5-Oxopyrrolidine-2-carboxylic acid (D-Pyroglutamic acid) is a cyclic derivative of D-glutamate. It can be used in the research of metabolic syndrome and end-stage renal disease.
  • Cyclic derivative of D-glutamate
  • Used in research of metabolic syndrome
  • Used in research of end-stage renal disease
Catalog No. 50-004-49465