Learn More
Medchemexpress LLC (R)-6-(1-hydroxy-1-methylethyl)-4-(2-fluorophenyl)-1-((E)-3-fluorostyryl)-1H-pyrazolo[3,4-c]pyridin | 425377-76-0 | 98.5% | 391.37 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY195055MG
Encompass_Preferred
TPA-023B is a high-affinity and orally active GABAA receptor α2/α3 subtype (Kis of 0.73 nM/2 nM) partial agonist and a α1 subtype (Ki of 1.8 nM) antagonist. TPA-023B has non-sedating anxiolytic-like properties.
  • High-affinity and orally active
  • GABAA receptor α2/α3 subtype partial agonist
  • GABAA receptor α1 subtype antagonist
  • Non-sedating anxiolytic-like properties
  • High affinity for α5 subtype (Ki of 1.1 nM) of human recombinant GABAA receptor
  • Over 1500-fold lower affinity for the α4- and α6 containing subtypes (Ki > 1000 nM)
  • Comparable affinity for native rat GABAA receptors in different regions of the CNS (Ki of 0.32-0.99 nM in cerebellum, spinal cord and frontal cortex)
  • Anxiolytic in rodent and primate models of anxiety
  • No significant effects in rodent or primate assays of ataxia and/or myorelaxation, even at high doses (10 mg/kg) with greater than 99% occupancy
Catalog No. 50-004-01834