Learn More
Medchemexpress LLC (R)-6-(4-chlorophenyl)-3-(4-(2-cyclopropyl-2-hydroxyethoxy)-3-methoxyphenyl)thieno[3,2-d]pyrimidin-4(3H)-one | 1197420-05-5 | 99.8% | 468.95 g/mol | C24H21ClN2O4S | 5MG

Supplier: Medchemexpress LLC HY124335MG
BMS-819881 is a research-use melanin-concentrating hormone receptor 1 (MCHR1) antagonist supplied as a small-quantity solid for in vitro and preclinical studies. The compound has molecular formula C24H21ClN2O4S, molecular weight 468.95 g/mol, and CAS 1197420-05-5. Storage and solubility guidance are provided by the supplier; the material is intended for laboratory research only and is not for clinical use.
- Molecular formula C24H21ClN2O4S.
- Molecular weight 468.95 g/mol.
- CAS registry number 1197420-05-5.
- Purity 99.8% as supplied.
- Available in small vial sizes suitable for solution preparation.
- Storage: powder stable at -20°C (3 years) or 4°C (2 years); in solvent store at -80°C (6 months) or -20°C (1 month).
- Solubility: ≥ 2.5 mg/mL in 10% DMSO/corn oil; clear solution at this concentration.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.