Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-amlodipine | 103129-81-3 | 99.45% | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYB0317D

Encompass_Preferred

(R)-Amlodipine is the R-enantiomer of amlodipine. It is for research use only.

  • Cas number: 103129-81-3
  • Molecular weight: 408.88
  • Formula: C20H25ClN2O5
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Target: Calcium channel
  • Pathway: Membrane transporter/ion channel; neuronal signaling
  • Storage: Powder -20°C 3 years; in solvent -80°C 6 months, -20°C 1 month
  • Solubility (in vitro): Dmso: 100 mg/mL (244.57 mM; requires ultrasonic)

Catalog No. 50-004-51000


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.