Learn More
Medchemexpress LLC (R)-Amlodipine-d4 | 1346616-97-4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0317DS1MG
Encompass_Preferred
(R)-Amlodipine-d4 is the deuterium labeled (R)-Amlodipine. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled (R)-Amlodipine
  • Incorporates stable heavy isotopes as tracers for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • For research use only
Catalog No. 50-004-06671