Learn More
Medchemexpress LLC (R)-azetidine-2-carboxylic acid | 7729-30-8 | MFCD00800731 | 99.9% | 101.10 g/mol | C4H7NO2 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0177555G
Encompass_Preferred
(R)-azetidine-2-carboxylic acid is the R-enantiomer of azetidine-2-carboxylic acid supplied as a white to off-white solid for research and chemical synthesis. It is used as a non-cleavable ADC linker and as an alkyl chain building block for PROTACs and other conjugation chemistries. Store at 4°C and protect from light.
  • High purity chiral building block for ADC and PROTAC linker synthesis.
  • Molecular formula C4H7NO2; molecular weight 101.10 g/mol.
  • Available in small-scale quantities, typically 5 g packaging.
  • White to off-white solid; store at 4°C and protect from light.
Catalog No. 50-002-03069