Learn More
Medchemexpress LLC (R)-Benzyl mandelate | 97415-09-3 | MFCD00674031 | 98.0% | 242.27 g/mol | C15H14O3 | 25 G

Supplier: Medchemexpress LLC 25GHYW020655
(R)-Benzyl mandelate is the R-enantiomer of benzyl mandelate, a chiral α-hydroxy ester used as a synthetic intermediate in pharmaceutical and organic synthesis. It is supplied for research use and commonly employed as a stereoselective building block in laboratory syntheses.
- Chiral α-hydroxy ester useful as a synthetic intermediate.
- High chemical purity (98.0%).
- Molecular formula C15H14O3 and molecular weight 242.27 g/mol.
- Available in lab-scale quantities (5 g-50 g).
- Suitable for stereoselective transformations and derivatizations.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.