Learn More
Medchemexpress LLC (R)-BI-2852 | 2375482-49-6 | 97.3% | 516.59 g/mol | C31H28N6O2 | 50MG

Supplier: Medchemexpress LLC HY126247B50MG
(R)-BI-2852 is the R-enantiomer of a KRAS switch I/II-pocket inhibitor supplied for research use as an experimental control. It preferentially binds active KRAS mutants and disrupts GEF, GAP, and effector interactions, producing antiproliferative effects in KRAS-mutant cells. Supplied as a solid with high enantiomeric purity and recommended storage under inert atmosphere.
- High enantiomeric purity (97.3%)
- Molecular formula C31H28N6O2
- Molecular weight 516.59 g/mol
- Soluble in DMSO (≈55 mg/mL)
- Available in multiple package sizes, including 50 mg
- Store solid at 4°C under nitrogen
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.