Learn More
Medchemexpress LLC HY-108250 5mg Medchemexpress, (R)-Bicalutamide CAS:113299-40-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1082505MG
Encompass_Preferred
Medchemexpress, HY-108250 5mg (R)-Bicalutamide CAS:113299-40-4 (R)-Bicalutamide is the (R)-enantiomer of Bicalutamide (HY-14249). (R)-Bicalutamide is an androgen receptor (AR) antagonist, with antineoplastic activity. (R)-Bicalutamide is widely used for the research of prostate cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0734