Learn More
Cayman Chemical R- Docosahexaenyl-2-Hyd 10mg
SDP
Supplier:  Cayman Chemical 900123010 MG
Encompass_Preferred
A homolog of DHEA, characterized by the addition of an (R)-β-methyl group at the terminal ethanolamine carbon
Catalog No. 50-289-3906