Learn More
Medchemexpress LLC (R)-Donepezil | 142698-19-9 | 99.9% | 379.49 | 1 ML

Supplier: Medchemexpress LLC HY13683810MM/1ML
(R)-Donepezil is the R-enantiomer of Donepezil and functions as a specific and potent acetylcholinesterase (AChE) inhibitor. It is characterized by its molecular formula C24H29NO3 and a molecular weight of 379.49.
- Specific and potent AChE inhibitor.
- Targets cholinesterase (ChE) in neuronal signaling pathways.
- Recommended storage for powder: -20°C for 3 years, 4°C for 2 years.
- Recommended storage for in-solvent form: -80°C for 6 months, -20°C for 1 month.
- Soluble in DMSO at 200 mg/mL with ultrasonic assistance.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.