Learn More
Medchemexpress LLC (R)-donepezil | 142698-19-9 | MFCD21396234 | 99.9% | C24H29NO3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13683810MG
Encompass_Preferred
(R)-Donepezil is the R-enantiomer of donepezil, a selective acetylcholinesterase (AChE) inhibitor supplied as a white to off-white solid for research use. It offers high enantiomeric purity and is intended for in vitro assays and preclinical in vivo studies investigating cholinergic modulation.
  • High chemical and enantiomeric purity.
  • White to off-white solid form, easy handling and storage.
  • Soluble in DMSO at high concentrations for in vitro use.
  • Includes validated in vivo formulation protocols for dosing.
  • Stable long-term under recommended cold storage conditions.
  • Suitable for AChE activity assays and pharmacology studies.
Catalog No. 50-002-60970