Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-Donepezil ((R)-E2020 free base) | 142698-19-9 | 99.91% ee.: 100.00% | 379.49 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY136838100MG

Encompass_Preferred

(R)-Donepezil is an R-enantiomer of Donepezil, a specific and potent Acetylcholinesterase (AChE) inhibitor. It is provided as a solid, white to off-white in appearance.

  • Specific and potent AChE inhibitor
  • R-enantiomer of Donepezil
  • Molecular weight: 379.49
  • Purity: 99.91%

Catalog No. 50-003-48699


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.