Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-eIF4A3-IN-2 | 2095484-82-3 | 602.70 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY4391350MG

Encompass_Preferred

(R)-eIF4A3-IN-2 is a less active enantiomer of eIF4A3-IN-2. It is a highly selective and noncompetitive eukaryotic initiation factor 4A-3 (eIF4A3) inhibitor with an IC50 of 110 nM. This compound targets eukaryotic initiation factor (eIF) and is involved in the cell cycle/DNA damage pathway.

  • Targets eukaryotic initiation factor (eIF)
  • Involved in cell cycle/DNA damage pathway
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 2 years, -20°C for 1 year
  • Solubility in DMSO: 50 mg/mL (need ultrasonic)
  • In vivo solubility: ≥ 2.5 mg/mL in 10% DMSO >> 40% PEG300 >> 5% Tween-80 >> 45% saline
  • In vivo solubility: ≥ 2.5 mg/mL in 10% DMSO >> 90% corn oil

Catalog No. 50-004-03886


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.