Learn More
Medchemexpress LLC (R)-Elsubrutinib (Synonyms: (R)-ABBV-105) | 1643570-23-3 | 98.0% | 297.35 | 1 MG

Supplier: Medchemexpress LLC HY109143B50MG
(R)-Elsubrutinib, also known as (R)-ABBV-105, is an inhibitor of Btk (Bruton's tyrosine kinase). It is intended for use in research related to various immune diseases and cancer. Potential applications include studies for conditions such as rheumatoid arthritis, psoriasis, ankylosing spondylitis, asthma, and systemic lupus erythematosus.
- Target: Btk inhibitor
- Applications:
- Immune diseases (e.g., rheumatoid arthritis, psoriasis, ankylosing spondylitis, asthma, systemic lupus erythematosus)
- Cancer
- Research use: For research purposes only; not sold to patients.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.