Learn More
Medchemexpress LLC (R)-Ethyl 5-(2,2-dimethyltetrahydro-2H-pyran-4-yl)-1H-indole-2-carboxylate | 2758661-69-5 | 301.38 | 5 G
SDP
Supplier:  Medchemexpress LLC HY23071A5G
Encompass_Preferred
(R)-Ethyl 5-(2,2-dimethyltetrahydro-2H-pyran-4-yl)-1H-indole-2-carboxylate is a drug intermediate for the synthesis of various active compounds.
  • Functions as a drug intermediate for synthesis.
  • Store at room temperature, keep dry and cool.
  • In solvent, stable at -80°C for 1 year and -20°C for 6 months.
Catalog No. 50-004-00873