Learn More
STA PHARMACEUTICAL US LLC (R)-Fmoc-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid | 10 g | CAS 204317-98-6 | MDL MFCD03788061
Supplier: STA PHARMACEUTICAL US LLC STAAA1017710GM
(R)-Fmoc-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid is a Amino Acid reagent (Subcategory: Aryl AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 204317-98-6
- MDL: MFCD03788061
- InChIKey: AGSTVRGPOSEJQQ-HSZRJFAPSA-N
- Molecular Weight: 399.446
- Molecular Formula: C25H21NO4
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29334990
- Country Of Origin: China
- IUPAC: (R)-2-(((9H-fluoren-9-yl)methoxy)carbonyl)-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid
- SMILES: O=C(O)[C@@H]1N(C(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O)CCC5=CC=CC=C51
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.