Learn More
Enzo Life Sciences R-(-)-Glucoraphanin . potassium salt (10mg). CAS: 21414-41-5
SDP
Supplier:  Enzo Life Sciences ALX350181M010
Encompass_Preferred
Modulator of hepatic cytochrome P450 and phase II conjugation activity. The enzyme myrosinase transforms R-(-)-glucoraphanin into R-Sulforaphane (ALX-350-230). Alternative name: 4-Methylsulfinylbutylglucosinate . K. Purity: ≥99% (HPLC). Long Term Storage: -20°C.
Catalog No. 50-200-8777