Learn More
Medchemexpress LLC (R)-(-)-Gossypol acetic acid | 866541-93-7 | C32H34O10 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15464A50MG
Encompass_Preferred
(R)-(-)-Gossypol acetic acid is the levorotatory isomer of the natural product Gossypol. It binds to Bcl-2, Mcl-1, and Bcl-xL proteins with high affinity, showing potent activity against head and neck squamous cell carcinomas (HNSCC) cell lines in vitro. It induces apoptosis in HNSCC tumor cells with functional p53 and also kills tumor cells with mutant p53 via a different mechanism.
  • Binds to Bcl-2, Mcl-1, and Bcl-xL proteins
  • More potent than (+)-Gossypol in inhibiting cell growth and inducing apoptosis
  • Exhibits greater growth inhibition relative to (±)-Gossypol than (+)-Gossypol in UM-SCC-6 and UM-SCC-14A cell lines
  • Shows potent activity against head and neck squamous cell carcinomas (HNSCC) cell lines in vitro
  • Induces apoptosis in HNSCC tumor cells with functional p53
  • Kills tumor cells with mutant p53 via a different mechanism
Catalog No. 50-003-76490