Learn More
TARGETMOL CHEMICALS INC R-IMPP 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T405810MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. R-IMPP is an inhibitor of PCSK9 translation. R-IMPP stimulates uptake of LDL-C in hepatoma cells by increasing LDL-R levels without altering levels of secreted transferrin. purity: 100%
Catalog No. 50-257-9982