Learn More
Medchemexpress LLC (R)-Isomucronulatol | 64474-51-7 | 5 MG

Supplier: Medchemexpress LLC HYN2495A5MG
(R)-Isomucronulatol is a natural flavonoid that can be isolated from the seeds of sphaerophysa salsula. This product is intended for research use only.
- Natural flavonoid
- Isolated from the seeds of sphaerophysa salsula
- For research use only
- Store between 2°C to 8°C
- Estimated purity of 99.94%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.