Learn More
TARGETMOL CHEMICALS INC R-JAK2 STAT3-IN-10a 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T72754L1MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (R)-JAK2/STAT3-IN-10a is the R-isomer of JAK2/STAT3-IN-1.JAK2/STAT3-IN-1 is a GP130 D1 structural domain inhibitor with antitumor activity that inhibits the phosphorylation of JAK2 and STAT3. The KD value of (R)-JAK2/STAT3-IN-1 on GP130 protein is 3.8 uM. Purity 98.34%
Catalog No. 50-288-0141