Learn More
Medchemexpress LLC (R)-JNJ-40418677 | 1146594-85-5 | 99.3% | 480.44 g/mol | C26H22F6O2 | 1 ML

Supplier: Medchemexpress LLC HY100604A10MM 1ML
(R)-enantiomer of a γ-secretase modulator supplied for laboratory research use. Provided as a solid or as a ready-to-use 10 mM solution in DMSO (1 mL). The high-purity compound is suitable for biochemical and cellular assays studying amyloid-β modulation; not for human or clinical use.
- High purity suitable for research: 99.3%.
- Available as powder and as a 10 mM solution in DMSO (1 mL).
- Molecular formula C26H22F6O2 and molecular weight 480.44 g/mol.
- Stable when stored as powder at -20°C or 4°C; follow recommended solvent storage conditions.
- Intended for in vitro and in vivo research applications requiring stereochemical specificity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.