Learn More
Medchemexpress LLC R-lercanidipine-d3 hydrochloride | 1217724-52-1 | MFCD28898581 | 99.9% | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0612DS10MG
Encompass_Preferred
(R)-Lercanidipine-d3 hydrochloride is the deuterium-labeled R-enantiomer of lercanidipine hydrochloride, supplied as an isotopically labeled reference standard for bioanalytical and pharmacokinetic studies. It is provided as a hydrochloride salt with high HPLC purity suitable for use as an internal standard in LC-MS assays.
  • Isotopically labeled (d3) R-enantiomer for use as an internal standard.
  • Hydrochloride salt with a molecular weight of 651.21.
  • High HPLC purity (99.94%) for reliable quantification.
  • CAS number 1217724-52-1 for unambiguous identification.
  • Intended for bioanalytical and pharmacokinetic assay development.
Catalog No. 50-002-98961