Learn More
Medchemexpress LLC (R)-Leucic acid (D-α-Hydroxyisocaproic acid) | 20312-37-2 | 98.0% | 132.16 | 5 G
SDP
Supplier:  Medchemexpress LLC HY302165G
Encompass_Preferred
(R)-Leucic acid (D-α-Hydroxyisocaproic acid) is an orally active D-isomer of the α-hydroxy analogue of Leucine. It is a metabolite of Lactobacillus and can promote intestinal fatty acid absorption by upregulating CD36 expression. (R)-Leucic acid can be used to study microbe-host interactions and the regulation of lipid metabolism by probiotics.
  • Orally active D-isomer
  • Metabolite of Lactobacillus
  • Promotes intestinal fatty acid absorption by upregulating CD36 expression
  • Useful for studying microbe-host interactions
  • Useful for studying the regulation of lipid metabolism by probiotics
Catalog No. 50-004-04930